EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Asymmetric alkynylation of aldehydes with propiolates without high reagent loading and any additives.
PMID 21552582
Abstract
The asymmetric alkynylation of aliphatic and aromatic aldehydes with propiolates was mediated by dialkylzinc and a novel prolinol catalyst without high reagent loading and any additives, such as Ti(Oi-Pr)(4), to give the corresponding γ-hydroxy-α,β-acetylenic esters with high enantiomeric excess of up to 95%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
186511
(S)-(+)-2-Pyrrolidinemethanol, 97%
C5H11NO
P51400
Propiolic acid, 95%
C3H2O2