EMAIL THIS PAGE TO A FRIEND
Journal of applied oral science : revista FOB
HOXB5 expression in oral squamous cell carcinoma.
PMID 21552713
Abstract
Human HOX genes encode transcription factors that act as master regulators of embryonic development. They are important in several processes such as cellular morphogenesis and differentiation. The HOXB5 gene in particular has been reported in some types of neoplasm, but not in oral cancer. The present study investigated the expression of HOXB5 in oral squamous cell carcinoma (SCC) and in non-tumoral adjacent tissues, focusing on verifying its possible role as a broad tumor-associated gene and its association with histopathological and clinical (TNM) characteristics. RT-PCR was performed to amplify HOXB5 mRNA in 15 OSCCs and adjacent non-tumoral epithelium. A possible association with TNM and histopathologic data was verified by the chi-square and post-hoc t-test. HOXB5 was amplified in 60% non-tumoral epithelium and in 93.3% carcinomas. No statistically significant differences were found regarding the HOXB5 mRNA expression and TNM or histological grade. HOXB5 is expressed in OSCCs and its role in cancer progression should be further investigated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CKOZFN10245 CompoZr® Knockout ZFN Kit, Human HOXB5 (NM_002147)