EMAIL THIS PAGE TO A FRIEND
Cancer chemotherapy and pharmacology
5-FU-induced neurotoxicity in cancer patients with profound DPD deficiency syndrome: a report of two cases.
PMID 21553285
Abstract
5-Fluorouracil (5-FU) is a mainstay for treating various solid tumours in adults, including digestive and head and neck cancers. 5-FU-related toxicities usually include haematological, digestive and cutaneous features. Additionally, 5-FU has been described as being potentially neurotoxic in patients, but these side effects are quite rare in clinical practice. Here, we report two cases of sudden and unpredictable drug-induced neurotoxicities that occurred in patients undergoing their first course of 5-FU-based chemotherapy. None of these patients had any previous neurological disorder history, and both were treated following standard regimen (LV-5-FU2 and TPF for patient 1 and 2, respectively). Neurotoxicity included drowsiness, acute confusion plus dysarthria for the first patient and seizure, confusion and signs of metabolic encephalopathy for the second one. In addition, typical 5-FU-related severe toxicities (e.g. neutropenia and mucosities) were observed. Both patients slowly recovered from these neurological toxicities under supportive treatment. It was assumed that overexposure to 5-FU could explain the severe toxicities encountered. To test this hypothesis, we retrospectively evaluated the dihydropyrimidine dehydrogenase (DPD) activity of these patients on a phenotypic basis. Evaluation of the uracil-to-di-hydrouracil (U/UH2) ratio in plasma revealed a profound DPD deficiency syndrome in both patients. These cases suggest that 5-FU standard dosage administration may lead to strong overexposure, responsible for the severe toxicities observed, including the neurological features. It implies that DPD deficiency can cause neurotoxicity in 5-FU-treated patients and advocates for the prospective screening of DPD deficiency before starting any 5-FU-containing chemotherapy so as to prevent such side effects in the future.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7628
Dihydrouracil
C4H6N2O2