EMAIL THIS PAGE TO A FRIEND
Circulation
Ca(2+)-related signaling and protein phosphorylation abnormalities play central roles in a new experimental model of electrical storm.
PMID 21555709
Abstract
Electrical storm (ES), characterized by recurrent ventricular tachycardia/fibrillation, typically occurs in implantable cardioverter-defibrillator patients and adversely affects prognosis. However, the underlying molecular basis is poorly understood. In the present study, we report a new experimental model featuring repetitive episodes of implantable cardioverter-defibrillator firing for recurrent ventricular fibrillation (VF), in which we assessed involvement of Ca(2+)-related protein alterations in ES. We studied 37 rabbits with complete atrioventricular block for ≈80 days, all with implantable cardioverter-defibrillator implantation. All rabbits showed long-QT and VF episodes. Fifty-three percent of rabbits developed ES (≥3 VF episodes per 24-hour period; 103±23 VF episodes per rabbit). Expression/phosphorylation of Ca(2+)-handling proteins was assessed in left ventricular tissues from rabbits with the following: ES; VF episodes but not ES (non-ES); and controls. Left ventricular end-diastolic diameter increased comparably in ES and non-ES rabbits, but contractile dysfunction was significantly greater in ES than in non-ES rabbits. ES rabbits showed striking hyperphosphorylation of Ca(2+)/calmodulin-dependent protein kinase II, prominent phospholamban dephosphorylation, and increased protein phosphatase 1 and 2A expression versus control and non-ES rabbits. Ryanodine receptors were similarly hyperphosphorylated at Ser2815 in ES and non-ES rabbits, but ryanodine receptor Ser2809 and L-type Ca(2+) channel α-subunit hyperphosphorylation were significantly greater in ES versus non-ES rabbits. To examine direct effects of repeated VF/defibrillation, VF was induced 10 times in control rabbits. Repeated VF tissues showed autophosphorylated Ca(2+)/calmodulin-dependent protein kinase II upregulation and phospholamban dephosphorylation like those of ES rabbit hearts. Continuous infusion of a calmodulin antagonist (W-7) to ES rabbits reduced Ca(2+)/calmodulin-dependent protein kinase II hyperphosphorylation, suppressed ventricular tachycardia/fibrillation, and rescued left ventricular dysfunction. ES causes Ca(2+)/calmodulin-dependent protein kinase II activation and phospholamban dephosphorylation, which can explain the vicious cycle of arrhythmia promotion and mechanical dysfunction that characterizes ES.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3281
N-(6-Aminohexyl)-5-chloro-1-naphthalenesulfonamide hydrochloride
C16H21ClN2O2S · HCl