PloS one
Irvalec inserts into the plasma membrane causing rapid loss of integrity and necrotic cell death in tumor cells.
PMID 21556352
Abstract
Irvalec is a marine-derived antitumor agent currently undergoing phase II clinical trials. In vitro, Irvalec induces a rapid loss of membrane integrity in tumor cells, accompanied of a significant Ca(2+) influx, perturbations of membrane conductivity, severe swelling and the formation of giant membranous vesicles. All these effects are not observed in Irvalec-resistant cells, or are significantly delayed by pretreating the cells with Zn(2+). Using fluorescent derivatives of Irvalec it was demonstrated that the compound rapidly interacts with the plasma membrane of tumor cells promoting lipid bilayer restructuration. Also, FRET experiments demonstrated that Irvalec molecules localize in the cell membrane close enough to each other as to suggest that the compound could self-organize, forming supramolecular structures that likely trigger cell death by necrosis through the disruption of membrane integrity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
12107C
Fetal Bovine Serum, USA origin, γ-irradiated by SER-TAIN® process, sterile-filtered, suitable for cell culture
12303C
Fetal Bovine Serum, USDA approved countries, sterile-filtered, cell culture tested
F1051
Fetal Bovine Serum, Canada origin, sterile-filtered, suitable for cell culture
12003C
Fetal Bovine Serum, Australia origin, USDA approved, sterile-filtered, suitable for cell culture
12007C
Fetal Bovine Serum, Australia origin, γ-irradiated, sterile-filtered, suitable for cell culture
12203C
Fetal Bovine Serum, New Zealand origin, sterile-filtered, suitable for cell culture
F0926
Fetal Bovine Serum, USDA approved, sterile-filtered, suitable for cell culture
12306C
Fetal Bovine Serum, non-USA origin, from USDA approved countries, Heat Inactivated, sterile-filtered, suitable for cell culture
12006C
Fetal Bovine Serum, Australia origin, USDA approved, Heat Inactivated, suitable for cell culture
12207C
Fetal Bovine Serum, New Zealand origin, cell culture tested
F9423
Fetal Bovine Serum, Australia origin, suitable for, USDA approved, sterile-filtered, suitable for cell culture, suitable for hybridoma
F3297
Fetal Bovine Serum, Australia origin, γ-irradiated, suitable for cell culture, suitable for hybridoma
12105C
Fetal Bovine Serum, Dialyzed by ultrafiltration against 0.15M NaCl, USA origin, sterile-filtered, cell culture tested
12103C
Fetal Bovine Serum, USA origin, cell culture tested
12106C
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture
F6765
Fetal Bovine Serum, USA origin, Charcoal Stripped, sterile-filtered, suitable for cell culture