EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Reusable bio-functionalized surfaces based on electrochemical desorption of benzenediazonium-grafted organic layers.
PMID 21559535
Abstract
We describe an electrochemical oxidative desorption of benzenediazonium-grafted organic layers and immobilized proteins on the layers from indium-tin-oxide electrode surfaces.