EMAIL THIS PAGE TO A FRIEND
Journal of the Medical Association of Thailand = Chotmaihet thangphaet
Ethion exposure and biological monitoring in vegetable farmers.
PMID 21560835
Abstract
To modify the method of analysis of urinary diethyl phosphate (DEP) in order to determine the relationship between atmospheric ethion concentration and urinary DEP concentration. A cross-sectional study was conducted by collecting atmospheric ethion in the breathing zone of 28 farmers following the NIOSH 5600 method. Urine samples were also collected to analyze urinary DEP concentrations by a modified method using a Gas Chromatography-Flame Photometric Detector (FPD). The average atmospheric ethion concentration in the breathing zone of farmers was 0.036 +/- 0.018 mg/m3. The average urinary DEP in pre-shift and post-shift was 0.030 +/- 0.06 and 0.851 +/- 1.80 mg/g of creatinine respectively. The average DEP during work shifts was 0.53 +/- 0.27, and ranged from 0.12 to 1.16 mg/g of creatinine. A high correlation coefficient (r = 0.645) was found between atmospheric ethion concentrations and urinary DEP concentrations at p < 0.001. The modified method provided a reliable result and the urinary DEP during work shifts was found to be a reliable biomarker of ethion exposure.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45477
Ethion, PESTANAL®, analytical standard
C9H22O4P2S4