EMAIL THIS PAGE TO A FRIEND
Nano letters
Enhancing oxide ion incorporation kinetics by nanoscale Yttria-doped ceria interlayers.
PMID 21563786
Abstract
Interlayering 17.5 nm of Yttria-doped ceria (YDC) thin films between bulk yttria-stabilized-zirconia electrolyte and a porous Pt cathode enhanced the performance of low-temperature solid oxide fuel cells. The added YDC interlayer (14.11% doped Y(2)O(3)) was fabricated by atomic layer deposition and reduced the cathode/electrolyte interfacial resistances while increasing the exchange current density j(0) by a factor of 4 at operating temperatures between 300-500 °C. Tafel plots and the fitted impedance data suggest that the charge transfer coefficient α of interlayered SOFCs was 1.25 times higher, and the electrode/interfacial activation energy was reduced from 0.85 to 0.76 eV.