EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Determination of fumaric and maleic acids with stacking analytes by transient moving chemical reaction boundary method in capillary electrophoresis.
PMID 21565348
Abstract
The paper presents an on-line transient moving chemical reaction boundary (MCRB) method for simply but efficiently stacking analytes in capillary electrophoresis (CE). The CE technique was developed for a rapid determination of fumaric and maleic acid. Based on the theory of MCRB, Effects of several important factors such as the pH and concentration of running buffer and the conditions of stacking analytes were investigated to acquire the optimum conditions. The optimized separations were carried out in a 20 mmol/L sulphate neutralized with ethylenediamine to pH 6.0 electrolytes using a capillary coated with poly (diallyldimethylammonium chloride) and direct UV detection at 214 nm. The optimized preconcentrations were carried out in 50 mmol/L borax (pH 9.0). The calibration curves were linear in the concentration range of 1.0×10⁻⁷-1.0×10⁻⁴ mol/L and 5.0×10⁻⁷-1.0×10⁻⁴ mol/L for fumaric and maleic acid with correlation coefficients higher than 0.9991. The detection limits were 5.34×10⁻⁸ mol/L for fumaric acid and 1.92×10⁻⁷ mol/L for maleic acid. This method was applied for determination of fumaric acid in apple juice and of fumaric and maleic acid in dl-malic, the recovery tests established for real samples were within the range 95-105%. This work provided a valid and simple approach to detect fumaric and maleic acid.