EMAIL THIS PAGE TO A FRIEND
Biomedical and environmental sciences : BES
Methylation of RAR-β2, RASSF1A, and CDKN2A genes induced by nickel subsulfide and nickel-carcinogenesis in rats.
PMID 21565688
Abstract
To investigate the expression variation of RAR-β2, RASSF1A, and CDKN2A gene in the process of nickel-induced carcinogenesis. Nickel subsulfide (Ni(3)S(2)) at dose of 10 mg was given to Wistar rats by intramuscular injection. The mRNA expression of the three genes in induced tumors and their lung metastasis were examined by Real-time PCR. The methylation status of the 5' region of these genes were detected by Quantitative Real-time methylation specific PCR. The mRNA expressions of the three genes both in muscle and lung tumor were decreased distinctly in comparison with normal tissue. But hypermethylation was found only in muscle tumor. These findings suggest that loss of function or decrease of RAR-β2, RASSF1A, and CDKN2A, as well as the hypermethylation of 5' region of these genes, are related with nickel exposure.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
343226
Nickel sulfide, 99.7% trace metals basis, −150 mesh
Ni3S2