EMAIL THIS PAGE TO A FRIEND
Small (Weinheim an der Bergstrasse, Germany)
Selective nanopatterning of protein via ion-induced focusing and its application to metal-enhanced fluorescence.
PMID 21567946
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F9636 IgG−FITC from human serum, ~20 mg/mL, buffered aqueous solution