EMAIL THIS PAGE TO A FRIEND
Future oncology (London, England)
Uroporphyrinogen decarboxylase: optimizing radiotherapy for head and neck cancer.
PMID 21568673
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0552
Tirapazamine, ≥98% (HPLC)
C7H6N4O2