EMAIL THIS PAGE TO A FRIEND
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine
Studies for the application of boron neutron capture therapy to the treatment of differentiated thyroid cancer.
PMID 21570311
Abstract
The aim of these studies was to evaluate the possibility of treating differentiated thyroid cancer by BNCT. These carcinomas are well controlled with surgery followed by therapy with (131)I; however, some patients do not respond to this treatment. BPA uptake was analyzed both in vitro and in nude mice implanted with cell lines of differentiated thyroid carcinoma. The boron intracellular concentration in the different cell lines and the biodistribution studies showed the selectivity of the BPA uptake by this kind of tumor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
17755
4-Borono-L-phenylalanine, ≥95.0% (HPLC)
C9H12BNO4