EMAIL THIS PAGE TO A FRIEND
Nanotechnology
Transparent nanostructured coatings with UV-shielding and superhydrophobicity properties.
PMID 21576801
Abstract
Visible light transparent, UV-shielding and superhydrophobic nanostructured coatings have been successfully fabricated through a facile layer-by-layer deposition of TiO(2) and SiO(2) nanoparticles. The coatings are composed of an underlying UV-shielding TiO(2) layer and a top fully covered protective SiO(2) layer. The resulting coatings can block 100% of UVB and UVC and almost 85% of UVA. The fabricated surfaces have contact angles exceeding 165° after coating with organic PTES (1H, 1H, 2H, 2H-perfluorooctyltriethoxysilane) molecules. The transparent superhydrophobic surfaces exhibit extremely strong UV stability. All coatings retain the initial UV-shielding and superhydrophobic properties even after exposure to 275 nm UV light with a light intensity of 75 mW cm(-2) for 12 h.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V1005
[deamino-Cys1, D-Arg8]-Vasopressin acetate salt hydrate, ≥95% (HPLC)
C46H64N14O12S2 · xC2H4O2 · yH2O