EMAIL THIS PAGE TO A FRIEND
Shock (Augusta, Ga.)
What's new in Shock, June 2011?
PMID 21577084
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E9658
Enalaprilat dihydrate, ≥98% (HPLC)
C18H24N2O5 ·2H2O