EMAIL THIS PAGE TO A FRIEND
Anesthesiology
Regulation of spinal substance p release by intrathecal calcium channel blockade.
PMID 21577088
Abstract
The authors investigated the role of different voltage-sensitive calcium channels expressed at presynaptic afferent terminals in substance P release and on nociceptive behavior evoked by intraplantar formalin by examining the effects of intrathecally delivered N- (ziconotide), T- (mibefradil), and L-type voltage-sensitive calcium channel blockers (diltiazem and verapamil). Rats received intrathecal pretreatment with saline or doses of morphine, ziconotide, mibefradil, diltiazem, or verapamil. The effect of these injections upon flinching evoked by intraplantar formalin (5%, 50 μl) was quantified. To assess substance P release, the incidence of neurokinin-1 receptor internalization in the ipsilateral and contralateral lamina I was determined in immunofluorescent-stained tissues. Intrathecal morphine (20 μg), ziconotide (0.3, 0.6, and 1 μg), mibefradil (100 μg, but not 50 μg), diltiazem (500 μg, but not 300 μg), and verapamil (200 μg, but not 50 and 100 μg) reduced paw flinching in phase 2 compared with vehicle control (P < 0.05), with no effect on phase 1. Ziconotide (0.3, 0.6, and 1 μg) and morphine (20 μg) significantly inhibited neurokinin-1 receptor internalization (P < 0.05), but mibefradil, diltiazem, and verapamil at the highest doses had no effect. These results emphasize the role in vivo of N-type but not T- and L-type voltage-sensitive calcium channel blockers in mediating the stimulus-evoked substance P release from small primary afferents and suggest that T- and L-type voltage-sensitive calcium channel blockers exert antihyperalgesic effects by an action on other populations of afferents or mechanisms involving postsynaptic excitability.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1182
ω-Conotoxin MVIIA, ≥95% (HPLC)
C102H172N36O32S7