EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Atom arrangement strategy for designing a turn-on 1H magnetic resonance probe: a dual activatable probe for multimodal detection of hypochlorite.
PMID 21584328
Abstract
The first dual activatable hypochlorite ((-)OCl)-sensing probe was developed, based on a new proof-of-concept design involving signal-activatable (1)H chemical probes using the triple-resonance NMR technique. The probe enabled fluorescence-(1)H MR dual turn-on detection of (-)OCl in solution and in crude tissue extracts.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D104809
Dihydrocoumarin, 99%
C9H8O2
W238104
Dihydrocoumarin, ≥99%, FCC, FG
C9H8O2