EMAIL THIS PAGE TO A FRIEND
The journal of physical chemistry. A
Pressure effect on the nonradiative process of thioflavin-T.
PMID 21585210
Abstract
Time-resolved emission techniques were employed to study the nonradiative process of thioflavin-T (ThT) in 1-propanol, 1-butanol, and 1-pentanol as a function of the hydrostatic pressure. Elevated hydrostatic pressure increases the alcohol viscosity, which in turn strongly influences the nonradiative rate of ThT. A diamond-anvil cell was used to increase the pressure up to 2.4 GPa. We found that the nonradiative rate constant, k(nr), decreases with pressure. We further found a remarkable linear correlation between a decrease in k(nr) (increase in the nonradiative lifetime, τ(nr)) and an increase in the solvent viscosity. The viscosity was varied by a factor of 1000 and k(nr) was measured at high pressures, at which the nonradiative rate constant of the molecules decreased from (7 ps)(-1) to (13 ns)(-1), (13 ps)(-1) to (17 ns)(-1) and (17 ps)(-1) to (15 ns)(-1) for 1-propanol, 1-butanol, and 1-pentanol, respectively. The viscosity-dependence of k(nr) is explained by the excited-state rotation rate of the two-ring systems, with respect to each other.