EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Identification of LY83583 as a specific inhibitor of Candida albicans MPS1 protein kinase.
PMID 21586273
Abstract
Candida albicans is the most common and virulent fungus causing candidiasis in various parts of the body and can be lethal to immunocompromised patients. All currently known antifungal therapies are drugs which cause serious side effects in the host. An inhibitor specific for fungus survival is an ideal therapeutic. C. albicans MPS1 (monopolar spindle 1) has been reported as a kinase essential to its survival. Because CaMps1p shares limited sequence homology with the human ortholog (hMps1p), we screened for a chemical inhibitor in anticipation of finding one with Candida specific cytotoxicity. In vitro screening using a recombinant catalytic domain of CaMps1p identified LY83583 (6-anilino-5,8-quinolinedione), known as a guanylate cyclase inhibitor, to be blocking CaMps1p kinase activity. In addition to its in vitro kinase inhibition, LY83583 reduced the growth rate of C. albicans. Finally, we compared the inhibitory activity on CaMps1p and hMps1p among inhibitors against those kinases. LY83583 showed specific inhibition for CaMps1p with no effect on hMps1p activity. Conversely, the CaMps1p activity was not affected by known hMps1p inhibitors. These findings suggest that CaMps1p may well be an ideal target molecule for antifungal therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A6563
6-Anilinoquinoline-5,8-quinone, ≥95% (TLC), solid
C15H10N2O2