EMAIL THIS PAGE TO A FRIEND
Acta crystallographica. Section E, Structure reports online
N-(4-Nitro-pheneth-yl)formamide.
PMID 21587938
Abstract
The title compound, C(9)H(10)N(2)O(3), was synthesized by direct N-formyl-ation of 4-nitro-phenethyl-amine hydro-chloride with formic acid and sodium formate in the absence of catalyst and solvent. In the crystal structure, mol-ecules are linked by inter-molecular N-H⋯O hydrogen-bond inter-actions into chains parallel to the a axis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
184802
4-Nitrophenethylamine hydrochloride, 95%
C8H10N2O2 · HCl