EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Organic-inorganic nanocomposites by placing conjugated polymers in intimate contact with quantum rods.
PMID 21590817
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
737887
Benzylphosphonic acid, 97%
C7H9O3P