EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Self-activation in de novo designed mimics of cell-penetrating peptides.
PMID 21591041
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N32407
Bicyclo[2.2.1]hept-2-ene, 99%
C7H10
225541
Diisopropyl azodicarboxylate, 98%
C8H14N2O4