EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Regulatory, integrative and comparative physiology
Electroacupuncture improves rectal distension-induced delay in solid gastric emptying in dogs.
PMID 21593428
Abstract
The aim of this study was to investigate the effects and mechanisms of electroacupuncture (EA) on rectal distension (RD)-induced delay in solid gastric emptying in dogs. Gastric emptying of solids was assessed in 12 dogs chronically implanted with a duodenal cannula by collecting samples at different time points from the cannula and measuring the dried weights of the samples. Bethanechol and atropine were used to qualitatively validate the method. In separate experiments, gastric emptying of solids was measured in a number of sessions: control, RD, RD + sham-EA, RD + EA of 6 mA, RD + EA of 3 mA, and RD + EA + naloxone. The method of gastric emptying by collecting and drying gastric chyme from the duodenal cannula was found to be accurate and reliable. Using the method, we found gastric emptying to be accelerated with bethanechol (70.01 ± 8.10% vs. 82.61 ± 4.15%, P = 0.04, vs. control) and delayed with atropine (4.31 ± 1.57%, P < 0.001, vs. control). RD substantially and significantly delayed gastric emptying. EA, but not sham-EA, attenuated delayed gastric emptying induced by RD (sham-EA: 48.79 ± 9.47% vs. EA: 74.28 ± 5.96%, P < 0.01). The effect was more potent with EA of 6 mA than EA of 3 mA and blocked by naloxone. EA is able to attenuate RD-induced delay in gastric emptying of solids, and this ameliorating effect may be mediated via the opioid pathway. EA may have a therapeutic potential for treating delayed gastric emptying attributed to lower gut distension.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C5259
Carbamyl-β-methylcholine chloride, ≥99% (TLC), crystalline
C7H17ClN2O2