EMAIL THIS PAGE TO A FRIEND
Giornale italiano di cardiologia (2006)
[The EMPHASIS-HF study].
PMID 21593949
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0410000
Canrenone, European Pharmacopoeia (EP) Reference Standard
C22H28O3