EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
Kinetic study of the reaction of OH with CH2I2.
PMID 21594268
Abstract
Flash photolysis (FP) coupled to resonance fluorescence (RF) was used to measure the absolute rate coefficients (k(1)) for the reaction of OH(X(2)Π) radicals with diiodomethane (CH(2)I(2)) over the temperature range 295-374 K. The experiments involved time-resolved RF detection of the OH (A(2)Σ(+)→X(2)Π transition at λ = 308 nm) following FP of the H(2)O/CH(2)I(2)/He mixtures. The OH(X(2)Π) radicals were produced by FP of H(2)O in the vacuum-UV at wavelengths λ > 120 nm. Decays of OH radicals in the presence of CH(2)I(2) are observed to be exponential, and the decay rates are found to be linearly dependent on the CH(2)I(2) concentration. The results are described by the Arrhenius expression k(1)(T) = (4.2 ± 0.5) × 10(-11) exp[-(670 ± 20)K/T] cm(3) molecule(-1) s(-1). The implications of the reported kinetic results for understanding the atmospheric chemistry of CH(2)I(2) are discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
158429
Diiodomethane, ReagentPlus®, 99%, contains copper as stabilizer
CH2I2