EMAIL THIS PAGE TO A FRIEND
Organic letters
Catalytic asymmetric vinylogous Mannich-type (AVM) reaction of nonactivated α-angelica lactone.
PMID 21595425
Abstract
A direct highly diastereo- and enantioselective asymmetric vinylogous Mannich-type (AVM) reaction of aldimines with nonactivated natural α-angelica lactone has been successfully developed. It was demonstrated that the nonactivated natural α-angelica lactone is a useful vinylogous nucleophile to give the chiral δ-amino γ,γ-disubstituted butenolide carbonyl derivatives. The N,N'-dioxide L2-Sc(III) complex is efficient toward the obtention of a range of corresponding products with adjacent quaternary and tertiary stereocenters in excellent dr and ee values.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A86406
α-Angelica lactone, 98%
C5H6O2
W329304
α-Angelica lactone, 98%, FG
C5H6O2