EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Temperature dependence of charge separation and recombination in porphyrin oligomer-fullerene donor-acceptor systems.
PMID 21595470