EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
HPLC-FLD methods to quantify chloroaluminum phthalocyanine in nanoparticles, plasma and tissue: application in pharmacokinetic and biodistribution studies.
PMID 21596512
Abstract
Analytical and bioanalytical methods of high-performance liquid chromatography with fluorescence detection (HPLC-FLD) were developed and validated for the determination of chloroaluminum phthalocyanine in different formulations of polymeric nanocapsules, plasma and livers of mice. Plasma and homogenized liver samples were extracted with ethyl acetate, and zinc phthalocyanine was used as internal standard. The results indicated that the methods were linear and selective for all matrices studied. Analysis of accuracy and precision showed adequate values, with variations lower than 10% in biological samples and lower than 2% in analytical samples. The recoveries were as high as 96% and 99% in the plasma and livers, respectively. The quantification limit of the analytical method was 1.12 ng/ml, and the limits of quantification of the bioanalytical method were 15 ng/ml and 75 ng/g for plasma and liver samples, respectively. The bioanalytical method developed was sensitive in the ranges of 15-100 ng/ml in plasma and 75-500 ng/g in liver samples and was applied to studies of biodistribution and pharmacokinetics of AlClPc.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
362530
Aluminum phthalocyanine chloride, Dye content ~85 %
C32H16AlClN8