EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Immunomodulatory effect of (--)-matairesinol in vivo and ex vivo.
PMID 21597179
Abstract
Matairesinol is one of the lignan compounds found in a variety of plant foodstuffs. We investigated the immunomodulatory effects of (-)-matairesinol in vivo and ex vivo by using mice. Although we found no significant differences in the IgG, IgA and IgM levels in the serum, the IgE level was strongly suppressed by the uptake of (-)-matairesinol in both intact and ovalbumin-immunized mice. The immunoglobulin produced by lymphocytes from the spleen was not activated by the intake of (-)-matairesinol. However, lymphocytes in such gut-associated lymphatic tissues as Peyer's patches and mesenteric lymph nodes were activated by the administration of (-)-matairesinol.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
40043
Matairesinol, ≥85% (HPLC)
C20H22O6
04157
Matairesinol, analytical standard
C20H22O6