EMAIL THIS PAGE TO A FRIEND
Current eye research
Human T lymphoblast cell line expresses FP receptor.
PMID 21599456
Abstract
To identify the presence of prostanoid FP receptor in human T lymphoblast (MOLT-3) cell line, which was known not to express FP receptor. Western blot analysis and mass spectrometry were performed on the cell lysates. The expression of FP receptor was detected by western blot analysis and mass spectrometry. MOLT-3 cell does express prostanoid FP receptor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
MOLT-3, 90021901