EMAIL THIS PAGE TO A FRIEND
Toxicology in vitro : an international journal published in association with BIBRA
p38 MAPK activation is required for esculetin-induced inhibition of vascular smooth muscle cells proliferation.
PMID 21600278
Abstract
The phenolic compound esculetin is known to inhibit the proliferation of vascular smooth muscle cells (VSMC). However, the signaling pathway by which esculetin mediates its molecular effects in VSMC remains to be identified. The present results suggest an unexpected role of the p38 MAPK signaling pathway in esculetin-induced inhibition of VSMC growth. Treatment of VSMC with esculetin resulted in significant growth inhibition and G1-phase cell-cycle arrest, which was followed by down-regulation of cyclins and cyclin-dependent kinase (CDK) expression. This G1-phase cell-cycle arrest was due to up-regulation of p21WAF1 expression. In addition, esculetin treatment activated p38 MAPK and ERK1/2. Pretreatment with SB203580, which is a p38 MAPK specific inhibitor, or expression of the dominant negative p38 MAPK (DN p38 MAPK) gene blocked esculetin-induced p38 MAPK activation and p21WAF1 expression. Finally, both the growth inhibition and the down-regulation of CDKs induced by esculetin were suppressed by either SB203580 or the DN p38 MAPK mutant gene. In conclusion, these results demonstrate that activation of p38 MAPK contributes to esculetin-induced p21WAF1 expression in VSMC by decreasing both the cyclin D1/CDK4 and cyclin E/CDK2 complexes. These novel results regarding the molecular mechanism of esculetin action suggest new preventive and therapeutic treatments for atherosclerosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
246573
6,7-Dihydroxycoumarin, 98%
C9H6O4