EMAIL THIS PAGE TO A FRIEND
Progress in nuclear magnetic resonance spectroscopy
MRI contrast agents based on dysprosium or holmium.
PMID 21600356
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
263028
Dysprosium, chips, 99.9% trace rare earth metals basis
Dy
261076
Dysprosium, ingot, 99.9% trace rare earth metals basis
Dy
GF48095185
Dysprosium, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
Dy
GF22672380
Dysprosium, wire reel, 25mm, diameter 1.0mm, hard, 99.9%
Dy
GF56425451
Dysprosium, wire reel, 100mm, diameter 0.5mm, hard, 99.9%
Dy
GF00748515
Dysprosium, wire reel, 50mm, diameter 0.5mm, hard, 99.9%
Dy
GF34048526
Dysprosium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%
Dy
GF11898096
Dysprosium, foil, 100x100mm, thickness 0.125mm, as rolled, 99%
Dy
GF69984769
Dysprosium, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
Dy
GF89687222
Dysprosium, foil, 25x25mm, thickness 0.1mm, as rolled, 99%
Dy
GF85958229
Dysprosium, foil, 25x25mm, thickness 0.25mm, as rolled, 99%
Dy
GF10903676
Dysprosium, foil, 50x50mm, thickness 0.1mm, as rolled, 99%
Dy
GF65566536
Dysprosium, foil, 50x50mm, thickness 0.25mm, as rolled, 99%
Dy
GF69010602
Dysprosium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Dy
GF04383592
Dysprosium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99.9%
Dy
GF13501358
Dysprosium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99.9%
Dy
GF89822065
Dysprosium, foil, not light tested, 5x5mm, thickness 0.005mm, as rolled, 99%
Dy
GF54698451
Dysprosium, powder, 1g, max. particle size 500 micron, 99.9%
Dy
GF87801723
Dysprosium, powder, 5g, max. particle size 500 micron, 99.9%
Dy
GF74506899
Dysprosium, rod, 100mm, diameter 6.35mm, cast, 99%
Dy
GF40435039
Dysprosium, rod, 50mm, diameter 2.0mm, cast, 99%
Dy
GF30196054
Dysprosium, rod, 50mm, diameter 6.35mm, cast, 99%
Dy
GF15989978
Dysprosium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%
Dy
GF99502836
Dysprosium, wire reel, 25mm, diameter 0.5mm, hard, 99.9%
Dy
GF56066171
Dysprosium, powder, 10g, max. particle size 500 micron, 99.9%
Dy
457957
Holmium, chips, 99.9% trace metals basis
Ho
693693
Holmium, foil, thickness 1 mm, 99.9% trace rare earth metals basis
Ho
755974
Holmium, dendritic pieces, purified by distillation, 99.99% trace rare earth metals basis
Ho
GF92551259
Holmium, rod, 50mm, diameter 3.8mm, 99%
Ho
GF12230154
Holmium, rod, 50mm, diameter 6.35mm, cast, 99%
Ho