EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Novel liquid-liquid-solid microextraction method with molecularly imprinted polymer-coated stainless steel fiber for aqueous sample pretreatment.
PMID 21600579
Abstract
A novel liquid-liquid-solid microextraction (LLSME) method was developed to overcome the well-known water-compatibility problem of molecularly imprinted polymers (MIPs). The enrichment factors with MIP-LLSME method were within 70-210 for trace chloroacetanilide herbicides under optimized extraction conditions. The method was characterized by simplicity, low solvent-consumption and high selectivity, and it was suitable for the one-step pretreatment of various aqueous samples such as river water and farm water.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
37887
Butachlor, PESTANAL®, analytical standard
C17H26ClNO2
34056
Propisochlor, PESTANAL®, analytical standard
C15H22ClNO2