EMAIL THIS PAGE TO A FRIEND
Fitoterapia
Bisindole alkaloids and secoiridoids from Alstonia macrophylla Wall. ex G. Don.
PMID 21601617
Abstract
The ethanolic extract from stems of a Thai medicinal plant, Alstonia macrophylla Wall. ex G. Don (Apocynaceae) showed a significant inhibitory effect on acetylcholinesterase (AChE) determined by using Ellman assay. Four compounds i.e., a bisindole alkaloid, macralstonine (1), a new bisindole alkaloid, thungfaine (2), a secoiridoid glycoside, sweroside (3) and a new secoiridoid glycoside, naresuanoside (4) were isolated. Compound 4 showed moderate AChE and butyrylcholinesterase (BChE) inhibitory effects. Interestingly, compound 4 inhibited cell growth on human androgen-sensitive prostate cancer cell line (LNCaP) but no effect on viability of human foreskin fibroblast cells (HF).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMB00083
Sweroside, ≥95% (LC/MS-ELSD)
C16H22O9