EMAIL THIS PAGE TO A FRIEND
Tetrahedron letters
Synthesis of C1-C20 and C21-C40 fragments of tetrafibricin.
PMID 21603053
Abstract
Efficient syntheses of suitably functionalized top and bottom fragments of tetrafibricin are described. The bottom fragment is prepared by two consecutive Kocienski-Julia couplings, while the top fragment synthesis features a dithiane alkylation and a Horner-Wadsworth-Emmons reaction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
375276
Di-tert-butyl-iminodicarboxylate, ≥96%
C10H19NO4