EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Highly enantioselective organocascade intermolecular iminium/enamine Michael addition on enals.
PMID 21603716
Abstract
An unprecedented intermolecular iminium/enamine Michael addition on enals has been developed by taking advantage of the high reactivity of vinyl sulfones. This powerful organocascade allows for the rapid construction of attractive synthons in high enantioselectivities (typically 99% ee).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V3700
Divinyl sulfone, contains ≤500 ppm hydroquinone as inhibitor, 97%
C4H6O2S