EMAIL THIS PAGE TO A FRIEND
Molecular biology reports
Cloning and functional characterization of a caffeic acid O-methyltransferase from Trigonella foenum-graecum L.
PMID 21604170
Abstract
A cDNA encoding an O-methyltransferase (namely FGCOMT1) was identified from the medicinal plant Trigonella foenum-graecum L. The FGCOMT1 enzyme is a functional caffeic acid O-methyltransferase (COMT) and is localized in the cytosol. Kinetic analysis indicated that FGCOMT1 protein exhibited the highest catalyzing efficiency towards 5-hydroxy ferulic acid and caffeic acid as substrates, but did not possess the abilities to methylate either quercetin or tricetin in vitro. Furthermore, transformation of Arabidopsis loss-of-function Atomt1 mutant with a FGCOMT1 cDNA partially complements accumulation of sinapoyl derivatives but did not function to produce the major methylated flavonol isorhamnetin in seeds. The results from this study indicated that FGCOMT1 is a COMT with substrate preference to monomeric lignin precursors but is not involved in the flavonoid methylation in T. foenum-graecum L.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
67563
3,4-Dihydroxy-5-methoxycinnamic acid, ≥95.0% (HPLC)
C10H10O5