EMAIL THIS PAGE TO A FRIEND
Mycoses
In vitro antifungal activity of coumarin extracted from Loeselia mexicana Brand.
PMID 21605187
Abstract
The bis-coumarin daphnoretin and its monomeric precursors scopoletin and umbelliferone were isolated for the first time from the aerial part of Loeselia mexicana Brand (a vegetal species used in Mexican traditional medicine) using chromatographic techniques. The structures of these compounds were determined by (1) H and (13) C NMR analyses. These coumarins were evaluated for in vitro antifungal activity. The three compounds tested showed significant antifungal activity.