EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Structure-function analysis of the tertiary bile acid TUDCA for the resolution of endoplasmic reticulum stress in intestinal epithelial cells.
PMID 21605547
Abstract
Inflammatory bowel diseases (IBD) are chronically relapsing and immune-mediated disorders of the gastrointestinal tract. Endoplasmic reticulum (ER) stress mechanisms in the epithelium have been demonstrated to be implemented into the pathogenesis of intestinal inflammation. Chemical chaperones have been demonstrated to exhibit beneficial effects in various diseases associated with ER stress mechanisms by prohibiting the unfolded protein response (UPR). In a structure-function analysis, we tested the potential of the conjugated bile salt sodium tauroursodeoxycholate (TUDCA), naturally present in the small bowel, to resolve ER stress in intestinal epithelial cells. TUDCA efficiently inhibited the expression of UPR dependent genes like GRP78 triggered by the ER stressor tunicamycin in the small intestinal epithelial cell line Mode-K. TUDCA inhibited upstream signaling events in all three branches of the UPR cascade and diminished binding of UPR activated transcription factors to the grp78 promoter. A structure-function analysis revealed that UDCA but not its conjugation partner taurine, known as a chemical chaperone, is responsible for the inhibition of GRP78 induction and that UDCA is 10 times more effective than its taurine conjugate. This inhibitory effect was confirmed in a cell free assay, where TUDCA and UDCA but not taurine effectively inhibited the aggregation of thermally denatured BSA. We conclude that TUDCA and UDCA are potent anti-aggregants for the resolution of ER stress in intestinal epithelial cells and should be considered as a potential drug target to resolve ER stress mechanisms underlying the pathology of IBD.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T0557
Sodium taurodeoxycholate hydrate, BioXtra, ≥97% (TLC)
C26H44NNaO6S · xH2O