EMAIL THIS PAGE TO A FRIEND
Journal of Lipid Research
Hyperglycemic Ins2AkitaLdlr?? mice show severely elevated lipid levels and increased atherosclerosis: a model of type 1 diabetic macrovascular disease.
PMID 21606463