EMAIL THIS PAGE TO A FRIEND
Surgical and Radiologic Anatomy : SRA
The Morphological characteristics of corticostriatal and thalamostriatal neurons and their intrastriatal terminals in rats.
PMID 21607670