EMAIL THIS PAGE TO A FRIEND
Indian journal of experimental biology
Comparative study of 6-APA production by free and agar immobilized bacteria in nutrient broth culture.
PMID 21614893
Abstract
In the present study different bacterial samples were isolated from soil of different places of Dibrugarh and screened for biotransformation ability to produce 6-Aminopenicillanic acid. Among ten isolated bacterial samples, three gram positive bacterial samples designated as AKDD-2, AKDD-4 and AKDD-6 showed the production of 6-APA from penicillin G. Assessment of production of 6-APA after incubation in penicillin G (2 mg/ml) by three different samples separately in free and agar immobilization state was done by HPLC analysis. Reusability of immobilized cells was found successful up to 14 days.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A70909
(+)-6-Aminopenicillanic acid, 96%
C8H12N2O3S