EMAIL THIS PAGE TO A FRIEND
Journal of pediatric surgery
Preoperative administration of Sudan III and successful treatment of persistent chylous ascites in a neonate.
PMID 21616268
Abstract
Congenital chylous ascites is a rare entity, and surgical treatment is confined to selected intractable cases. We report 2 cases of refractory congenital chylous ascites successfully treated with preoperative administration of lipophilic dye (Sudan III) followed by abdominal systematic surgical exploration, cauterization, and fibrin glue application.