EMAIL THIS PAGE TO A FRIEND
American journal of botany
New primers for single-copy nuclear-encoded chloroplast-expressed glutamine synthetase (ncpGS) in Oxalidaceae.
PMID 21616835
Abstract
New primers were developed for the nuclear marker glutamine synthetase (ncpGS) in Oxalidaceae. • New forward and reverse primers were designed and tested across a wide range of Oxalidaceae. Selected taxa were sequenced to confirm homology. Potential for phylogenetic study was assessed by comparing sequenced taxa with commonly used nuclear and plastid markers. • Four out of five Oxalidaceae genera and all tested Oxalis spp. amplified successfully. Sequencing confirmed homology of the amplicon. Parsimony analysis of ncpGS showed that it is a promising candidate for future phylogenetic work in Oxalidaceae.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
NA1020
GenElute PCR Clean-Up Kit, sufficient for 70 purifications