EMAIL THIS PAGE TO A FRIEND
EMBO reports
A member of the ETS family, EHF, and the ATPase RUVBL1 inhibit p53-mediated apoptosis.
PMID 21617703
Abstract
Tumour cells are known to be dependent on, or 'addicted to', not only oncogenes, but also some non-oncogenes. However, the mechanisms by which tumour cells are addicted to these genes have not been fully explained. Here, we show that overexpression of a member of the ETS family, EHF, is required for the survival of colon tumour cells that contain wild-type p53. We found that EHF directly activates the transcription of RUVBL1, an ATPase associated with chromatin-remodelling complexes. RUVBL1 blocks p53-mediated apoptosis by repressing the expression of p53 and its target genes. Moreover, we found that RUVBL1 represses p53 transcription by binding to the p53 promoter, interfering with RNF20/hBRE1-mediated histone H2B monoubiquitination and promoting PAF1-mediated histone H3K9 trimethylation. These results indicate that EHF-mediated RUVBL1 expression allows colon tumour cells to avoid p53-mediated apoptosis. Thus, EHF and RUVBL1 might be promising molecular targets for the treatment of colon tumours.