EMAIL THIS PAGE TO A FRIEND
Advances in experimental medicine and biology
GM3 Upregulation of matrix metalloproteinase-9 possibly through PI3K, AKT, RICTOR, RHOGDI-2, and TNF-A pathways in mouse melanoma B16 cells.
PMID 21618116