EMAIL THIS PAGE TO A FRIEND
Medical hypotheses
Can bisphenol A diglycidyl ether (BADGE) administration prevent steroid-induced femoral head osteonecrosis in the early stage?
PMID 21620573
Abstract
Steroid-induced osteonecrosis of the femoral head (ONFH) is associated with increase of intraosseous pressure caused by elevating of adipogenesis and fat cell hypertrophy in the bone marrow, which subsequently decreases the blood flow in the femoral head and finally resulting in bone ischemia. The early femoral head-preserving method has mainly focused on the conventional core decompression procedure. However, it only achieves a slight decrease in intra-medullary pressure with limited clinical outcome. The crucial point in prevention is to achieve a thorough decompression of intra-medullary pressure and improvement of microcirculation of the femoral head. Bisphenol-A-diglycidyl ether (BADGE), an antagonism of PPAR-γ(Peroxisome proliferator-activated receptor gamma), has been shown to successfully reverse bone marrow adipogenesis and fat cell hypertrophy, enhances proliferation of osteoblasts, inhibit osteoclastogenesis. Therefore, we hypothesized that BADGE administration may be an appropriate novel method for the prevention of early stage steroid-induced ONFH.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3415
Bisphenol A diglycidyl ether
C21H24O4
15138
Bisphenol A diglycidyl ether, analytical standard
C21H24O4