EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Two new acridone alkaloids from Glycosmis macrantha.
PMID 21623311
Abstract
Extraction and chromatographic separation of the extracts of dried stem barks of Glycosmis macrantha lead to isolation of two new acridone alkaloids, macranthanine and 7-hydroxynoracronycine, and a known acridone, atalaphyllidine. The structures of these alkaloids were determined by detailed spectral analysis and also by comparison with reported data.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
150215
9(10H)-Acridanone, 99%
C13H9NO