EMAIL THIS PAGE TO A FRIEND
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment
Concentrations of bisphenol A in the composite food samples from the 2008 Canadian total diet study in Quebec City and dietary intake estimates.
PMID 21623504
Abstract
A total of 154 food composite samples from the 2008 total diet study in Quebec City were analysed for bisphenol A (BPA), and BPA was detected in less than half (36%, or 55 samples) of the samples tested. High concentrations of BPA were found mostly in the composite samples containing canned foods, with the highest BPA level being observed in canned fish (106 ng g(-1)), followed by canned corn (83.7 ng g(-1)), canned soups (22.2-44.4 ng g(-1)), canned baked beans (23.5 ng g(-1)), canned peas (16.8 ng g(-1)), canned evaporated milk (15.3 ng g(-1)), and canned luncheon meats (10.5 ng g(-1)). BPA levels in baby food composite samples were low, with 2.75 ng g(-1) in canned liquid infant formula, and 0.84-2.46 ng g(-1) in jarred baby foods. BPA was also detected in some foods that are not canned or in jars, such as yeast (8.52 ng g(-1)), baking powder (0.64 ng g(-1)), some cheeses (0.68-2.24 ng g(-1)), breads and some cereals (0.40-1.73 ng g(-1)), and fast foods (1.1-10.9 ng g(-1)). Dietary intakes of BPA were low for all age-sex groups, with 0.17-0.33 µg kg(-1) body weight day(-1) for infants, 0.082-0.23 µg kg(-1) body weight day(-1) for children aged from 1 to 19 years, and 0.052-0.081 µg kg(-1) body weight day(-1) for adults, well below the established regulatory limits. BPA intakes from 19 of the 55 samples account for more than 95% of the total dietary intakes, and most of the 19 samples were either canned or in jars. Intakes of BPA from non-canned foods are low.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
44010
Dodecane, analytical standard
C12H26
D221104
Dodecane, ReagentPlus®, ≥99%
C12H26