EMAIL THIS PAGE TO A FRIEND
Arquivos de neuro-psiquiatria
Autosomal recessive spastic ataxia of Charlevoix-Saguenay (ARSACS): typical clinical and neuroimaging features in a Brazilian family.
PMID 21625752
Abstract
Autosomal recessive spastic ataxia of Charlevoix-Saguenay (ARSACS) is a neurodegenerative disorder characterized by late-infantile onset spastic ataxia and other neurological features. ARSACS has a high prevalence in northeastern Quebec, Canada. Several ARSACS cases have been reported outside Canada in recent decades. This is the first report of typical clinical and neuroimaging features in a Brazilian family with probable diagnosis of ARSACS.